L-Chicoric acid ≥98.0%

Leverantör: TCI

Synonyms: Chicoric acid, (2R,3R)-2,3-Bis[[(E)-3-(3,4-dihydroxyphenyl)acryloyl]oxy]succinic acid

C3450-10MG C3450-50MG
TCIAC3450-10MGEA 1100 SEK
TCIAC3450-10MG TCIAC3450-50MG
L-Chicoric acid ≥98.0%
L-Chicoric acid
Formel: C₂₂H₁₈O₁₂
Smältpunkt: 206°C
Storage Temperature: Kylskåp
MDL Number: MFCD22683653
CAS nummer: 70831-56-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4eea27a5dde7424f01ac8744140473f63c8111f&vid=b4eea27a5dde7424f01ac8744140473f63c8111f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -360 to -400 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR