Calcium(II) bis(nonafluorobutanesulphonyl)imide ≥95.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
Bis(nonafluorobutanesulphonyl)imide Calcium(II) salt
C3447-1G
C3447-5G
TCIAC3447-1GEA
775
SEK
TCIAC3447-1G
TCIAC3447-5G
Calcium(II) bis(nonafluorobutanesulphonyl)imide ≥95.0%
Calcium(II) bis(nonafluorobutanesulphonyl)imide
Formel:
C₁₆CaF₃₆N₂O₈S₄ MW: 1200,44 g/mol |
CAS nummer:
689282-13-1 UN: 1759 ADR: 8,II |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Ion exchange titration) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...