3-Chloro-6-phenylpyridazine ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 3-Phenyl-6-chloropyridazine

C3440-1G C3440-5G
TCIAC3440-1GEA 1125 SEK
TCIAC3440-1G TCIAC3440-5G
3-Chloro-6-phenylpyridazine ≥98.0%
3-Chloro-6-phenylpyridazine
Formel: C₁₀H₇ClN₂
MW: 190,6300 g/mol
MDL Number: MFCD00075253
CAS nummer: 20375-65-9
EINECS: 243-772-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aae4fa6fe34774344df0d2f990f4e6e711c1e3de&vid=aae4fa6fe34774344df0d2f990f4e6e711c1e3de&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 157.0 to 161.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR