Uretan ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: Etylkarbamat
44804.18 44804.30
44804.18EA 990 SEK
44804.18 44804.30
Uretan ≥98%
Uretan
Purity: 98% CAS No.: 51-79-6 Molecular Formula: C3H7NO2 Formula Weight: 89.09 Flash Point: 92°(198°F) Physical Form: Crystalline Melting Point: 48-50° Boiling Point: 182-184° Beilstein Reference: 635810 Merck Reference: 14,9877 MDL No.: MFCD00007966
Formel: C₃H₇NO₂
MW: 89,09 g/mol
Kokpunkt: 182…184 °C
Smältpunkt: 48…50 °C
Densitet: 1,11 g/cm³ (20 °C)
Flampunkt: >92 °C (198 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00007966
CAS nummer: 51-79-6
EINECS: 200-123-1
UN: 2757
ADR: 6.1,II
Merck Index: 14,9877