2-Chloro-10-(3-chloropropyl)-10H-phenothiazine ≥98.0%
Leverantör: TCI EUROPE
Varning
C3394-250MG C3394-1G
TCIAC3394-250MGEA 2700 SEK
TCIAC3394-250MG TCIAC3394-1G
2-Chloro-10-(3-chloropropyl)-10H-phenothiazine ≥98.0%
2-Chloro-10-(3-chloropropyl)-10H-phenothiazine
Formel: C₁₅H₁₃Cl₂NS
MW: 310,24 g/mol
Kokpunkt: 135°C
MDL Number: MFCD01247123
CAS nummer: 2765-59-5
EINECS: 220-438-8
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %