Cryptotanshinone ≥97.0%

Leverantör: TCI EUROPE
Fara

Synonyms: (R)-1,2,6,7,8,9-Hexahydro-1,6,6-trimethylphenanthro[1,2-b]furan-10,11-dione, (R)-1,2,6,7,8,9-Hexahydro-1,6,6-trimethyl-phenanthro(1,2-b)furan-10,11-dione, Tanshinone C

C3363-25MG C3363-100MG
TCIAC3363-25MGEA 335 SEK
TCIAC3363-25MG TCIAC3363-100MG
Cryptotanshinone ≥97.0%
Cryptotanshinone
Formel: C₁₉H₂₀O₃
MW: 296,37 g/mol
Smältpunkt: 192°C
Storage Temperature: Kylskåp
MDL Number: MFCD07636810
CAS nummer: 35825-57-1
UN: 1544
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0510b952b0f23b3e5477fc8bad75b7c45160c24f&vid=0510b952b0f23b3e5477fc8bad75b7c45160c24f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Orange to Brown powder to crystal
Purity (HPLC) min. 97.0 area%
Specific rotation [a]20/D -28.0 to -32.0 deg (C=0.5,MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR