2-Chloro-9H-carbazole ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
C3362-1G
C3362-5G
TCIAC3362-1GEA
430
SEK
TCIAC3362-1G
TCIAC3362-5G
2-Chloro-9H-carbazole ≥98.0%
2-Chloro-9H-carbazole
Formel:
C₁₂H₈ClN MW: 201,65 g/mol |
MDL Number:
MFCD01444890 CAS nummer: 10537-08-3 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 243.0 to 247.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...