5-Chloro-2-methylbenzonitrile ≥98.0%

Leverantör: TCI EUROPE
Varning

C3339-1G C3339-5G
TCIAC3339-1GEA 310 SEK
TCIAC3339-1G TCIAC3339-5G
5-Chloro-2-methylbenzonitrile ≥98.0%
5-Chloro-2-methylbenzonitrile
Formel: C₈H₆ClN
MW: 151,5929 g/mol
MDL Number: MFCD00045599
CAS nummer: 50712-70-4

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 43.0 to 47.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR