3-Chloro-p-cresol ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
3-Chloro-4-methylphenol, 1-Chloro-3-hydroxy-6-methylbenzene
C3333-5G
C3333-25G
TCIAC3333-5GEA
525
SEK
TCIAC3333-5G
TCIAC3333-25G
3-Chloro-p-cresol ≥98.0%
3-Chloro-p-cresol
Formel:
C₇H₇ClO MW: 142,58 g/mol Kokpunkt: 228°C |
MDL Number:
MFCD00060319 CAS nummer: 615-62-3 EINECS: 210-439-1 UN: 3437 ADR: 6.1,II |
Specifikation testresultat
Appearance | Light yellow to Amber to Dark green powder to lump |
Purity (GC) | min. 98.0 % |
Melting point | 51.0 to 55.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...