Chlorobis[dicyclohexyl(phenyl)phosphino](o-tolyl)nickel(II)
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
Jamison Catalyst
C3325-100MG
C3325-500MG
TCIAC3325-100MGEA
975
SEK
TCIAC3325-100MG
TCIAC3325-500MG
Chlorobis[dicyclohexyl(phenyl)phosphino](o-tolyl)nickel(II)
Chlorobis[dicyclohexyl(phenyl)phosphino](o-tolyl)nickel(II)
Formel:
C₄₃H₆₁ClNiP₂ MW: 734,05 g/mol Smältpunkt: 150°C |
MDL Number:
MFCD27978415 CAS nummer: 1419179-26-2 |
Specifikation testresultat
Appearance | Light yellow to Yellow to Orange powder to crystal |
Elemental analysis (Carbon) | 67.00 to 74.00 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...