N-(4-Chloro-3-cyano-7-ethoxy-6-quinolinyl)acetamide ≥98.0% (by HPLC, total nitrogen)

Leverantör: TCI EUROPE
Varning

C3315-1G C3315-5G
TCIAC3315-1GEA 885 SEK
TCIAC3315-1G TCIAC3315-5G
N-(4-Chloro-3-cyano-7-ethoxy-6-quinolinyl)acetamide ≥98.0% (by HPLC, total nitrogen)
N-(4-Chloro-3-cyano-7-ethoxy-6-quinolinyl)acetamide
Formel: C₁₄H₁₂ClN₃O₂
MW: 289,72 g/mol
Smältpunkt: 250°C
MDL Number: MFCD09263704
CAS nummer: 848133-76-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db7678b52b074fd5003e1f5e3cae00bf847a16fa&vid=db7678b52b074fd5003e1f5e3cae00bf847a16fa&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR