λ-Carrageenan
Leverantör: TCI
Synonyms: lambda-Carrageenan (High-viscosity)
C3313-25G C3313-500G
TCIAC3313-25GEA 805 SEK
TCIAC3313-25G TCIAC3313-500G
λ-Carrageenan
lambda-Carrageenan
MDL Number: MFCD00151513
CAS nummer: 9064-57-7
EINECS: 232-953-5
Merck Index: 14,01864
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85a83389f1aeb190b2b8b2882b8fa02081030c4e&vid=85a83389f1aeb190b2b8b2882b8fa02081030c4e&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Gray to Brown powder to crystal
Drying loss max. 15.0 %
Viscosity min. 70 mPa-s (0.3 %, H2O, 25 deg-C)
Ash Content max. 30.0 %