2'-Chloro-5'-fluoroacetophenone ≥98.0%
Leverantör: TCI EUROPE
Varning
C3297-1G C3297-5G
TCIAC3297-1GEA 430 SEK
TCIAC3297-1G TCIAC3297-5G
2'-Chloro-5'-fluoroacetophenone ≥98.0%
2'-Chloro-5'-fluoroacetophenone
Formel: C₈H₆ClFO
MW: 172,59 g/mol
Kokpunkt: 82°C
MDL Number: MFCD00042574
CAS nummer: 2965-16-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d86af3b98c9484bcaffd28f9a5f11451243dfc5c&vid=d86af3b98c9484bcaffd28f9a5f11451243dfc5c&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %