4-Chloro-2-(trifluoromethyl)benzoic acid ≥98.0% (by GC, titration analysis)

Leverantör: TCI
Varning

Synonyms: 4-Chloro-α,α,α-trifluoro-o-toluic acid

C3285-1G C3285-5G
TCIAC3285-1GEA 310 SEK
TCIAC3285-1G TCIAC3285-5G
4-Chloro-2-(trifluoromethyl)benzoic acid ≥98.0% (by GC, titration analysis)
4-Chloro-2-(trifluoromethyl)benzoic acid
Formel: C₈H₄ClF₃O₂
MW: 224.57 g/mol
MDL Number: MFCD03094162
CAS nummer: 142994-09-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2acaab7478d3d3808680a01b1057d96f0716fe1a&vid=2acaab7478d3d3808680a01b1057d96f0716fe1a&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 108.0 to 112.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR