4'-Cyano-4-dodecyloxybiphenyl ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4'-(Dodecyloxy)-[1,1'-biphenyl]-4-carbonitrile, 4'-Dodecyloxy-4-biphenylcarbonitrile, 12OCB
C3270-1G
C3270-5G
TCIAC3270-1GEA
895
SEK
TCIAC3270-1G
TCIAC3270-5G
4'-Cyano-4-dodecyloxybiphenyl ≥98.0%
4'-Cyano-4-dodecyloxybiphenyl
Formel:
C₂₅H₃₃NO MW: 363,55 g/mol |
MDL Number:
MFCD01941019 CAS nummer: 57125-50-5 EINECS: 260-575-0 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Mesomorphic range | 84.0 to 92.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...