4-Carboxy-2-nitrophenylboronic acid (contains varying amounts of Anhydride)

Leverantör: TCI EUROPE
Varning

Synonyms: 4-Carboxy-2-nitrobenzeneboronic acid

C3265-1G C3265-5G
TCIAC3265-1GEA 405 SEK
TCIAC3265-1G TCIAC3265-5G
4-Carboxy-2-nitrophenylboronic acid (contains varying amounts of Anhydride)
4-Carboxy-2-nitrophenylboronic acid
Formel: C₇H₆BNO₆
MW: 210,94 g/mol
Smältpunkt: 261°C
MDL Number: MFCD00448780
CAS nummer: 85107-54-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85ed86b1e096acc67b3fab28f0fd12f6e3168392&vid=85ed86b1e096acc67b3fab28f0fd12f6e3168392&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 97.0 to 110.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR