Chloramphenicol Palmitate ≥97.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: D-(-)-threo-2-Dichloroacetamido-1-(4-nitrophenyl)-1,3-propanediol Palmitate, 2,2-Dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-nitrophenyl)-2-propyl]acetamide Palmitate

C3258-200MG C3258-1G C3258-5G
TCIAC3258-200MGEA 270 SEK
TCIAC3258-200MG TCIAC3258-1G TCIAC3258-5G
Chloramphenicol Palmitate ≥97.0% (by HPLC, titration analysis)
Chloramphenicol Palmitate
Formel: C₂₇H₄₂Cl₂N₂O₆
MW: 561,54 g/mol
MDL Number: MFCD00083597
CAS nummer: 530-43-8
EINECS: 208-477-9
Merck Index: 14,02077

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Argentmetric Titration) min. 97.0 %
Melting point 91.0 to 96.0 deg-C
Specific rotation [a]20/D +20.5 to +25.4 deg (C=2, EtOH (99.5))

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR