Carboxymethyldextran sodium salt, M.W. ~ 40000

Leverantör: TCI

C3251-1G C3251-5G
TCIAC3251-1GEA 470 SEK
TCIAC3251-1G TCIAC3251-5G
Carboxymethyldextran sodium salt, M.W. ~ 40000
Carboxymethyldextran sodium salt
MDL Number: MFCD00146477
CAS nummer: 39422-83-8

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Content (Sodium) 6.5 to 9.5 % (calcd.on dried substance)
Drying loss max. 8.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR