(S)-1-(Chloroacetyl)-2-pyrrolidinecarbonitrile ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: (S)-1-(2-Chloroacetyl)pyrrolidine-2-carbonitrile, (S)-1-(Chloroacetyl)-2-cyanopyrrolidine

C3202-25G C3202-5G
TCIAC3202-25GEA 3875 SEK
TCIAC3202-25G TCIAC3202-5G
(S)-1-(Chloroacetyl)-2-pyrrolidinecarbonitrile ≥98.0%
(S)-1-(Chloroacetyl)-2-pyrrolidinecarbonitrile
Formel: C₇H₉ClN₂O
MW: 172,61 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD08689902
CAS nummer: 207557-35-5

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 62.0 to 66.0 deg-C
Specific rotation [a]20/D -150.0 to -160.0 deg (C=1, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR