3-(1H-Indol-3-yl)-3-oxopropanenitrile ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
3-(Cyanoacetyl)indole
C3197-1G
C3197-5G
TCIAC3197-1GEA
1425
SEK
TCIAC3197-1G
TCIAC3197-5G
3-(1H-Indol-3-yl)-3-oxopropanenitrile ≥98.0%
3-(1H-Indol-3-yl)-3-oxopropanenitrile
Formel:
C₁₁H₈N₂O MW: 184,2 g/mol |
MDL Number:
MFCD04610443 CAS nummer: 20356-45-0 UN: 3439 ADR: 6.1,III |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...