2-Chloro-6-fluoropyridine ≥98.0%
Leverantör: TCI
|
Fara
|
C3186-5G
C3186-25G
TCIAC3186-5GEA
415
SEK
TCIAC3186-5G
TCIAC3186-25G
2-Chloro-6-fluoropyridine ≥98.0%
2-Chloro-6-fluoropyridine
Formel:
C₅H₃ClFN MW: 131.54 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD00077483 CAS nummer: 20885-12-5 |
Specifikation testresultat
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Melting point | 31.0 to 35.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...