2-Cyclohepten-1-one ≥85.0%
Leverantör: TCI
Synonyms: Cyclohept-2-enone
C3180-1G C3180-5G
TCIAC3180-1GEA 935 SEK
TCIAC3180-1G TCIAC3180-5G
2-Cyclohepten-1-one ≥85.0%
2-Cyclohepten-1-one
Formel: C₇H₁₀O
MW: 110.16 g/mol
Kokpunkt: 183°C
Flampunkt: 68°C
Storage Temperature: Kylskåp
MDL Number: MFCD00004157
CAS nummer: 1121-66-0
EINECS: 214-334-1
Order Now
Specifikation testresultat
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 85.0 %