4-Chloro-3-nitrobenzamide ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
C3176-1G
C3176-5G
TCIAC3176-1GEA
510
SEK
TCIAC3176-1G
TCIAC3176-5G
4-Chloro-3-nitrobenzamide ≥98.0%
4-Chloro-3-nitrobenzamide
Formel:
C₇H₅ClN₂O₃ |
MDL Number:
MFCD00017134 CAS nummer: 16588-06-0 EINECS: 240-644-1 |
Specifikation testresultat
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 152.0 to 156.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...