2-Chloro-4-phenylquinazoline ≥98.0%
Leverantör: TCI
C3173-1G
C3173-5G
TCIAC3173-1GEA
615
SEK
TCIAC3173-1G
TCIAC3173-5G
2-Chloro-4-phenylquinazoline ≥98.0%
2-Chloro-4-phenylquinazoline
Formel:
C₁₄H₉ClN₂ |
MDL Number:
MFCD01152724 CAS nummer: 29874-83-7 |
|
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 113.0 to 117.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...