1-(4-Chlorophenyl)cyclobutanecarbonitrile ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1-(4-Chlorophenyl)-1-cyclobutanecarbonitrile
C3161-5G
C3161-25G
TCIAC3161-5GEA
1450
SEK
TCIAC3161-5G
TCIAC3161-25G
1-(4-Chlorophenyl)cyclobutanecarbonitrile ≥98.0%
1-(4-Chlorophenyl)cyclobutanecarbonitrile
Formel:
C₁₁H₁₀ClN MW: 191,6568 g/mol Kokpunkt: 112°C |
MDL Number:
MFCD00065239 CAS nummer: 28049-61-8 EINECS: 248-799-7 |
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...