2-Chloro-5-fluoro-3-methylpyridine ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 2-Chloro-5-fluoro-3-picoline

C3144-1G C3144-5G
TCIAC3144-1GEA 535 SEK
TCIAC3144-1G TCIAC3144-5G
2-Chloro-5-fluoro-3-methylpyridine ≥98.0%
2-Chloro-5-fluoro-3-methylpyridine
Formel: C₆H₅ClFN
MW: 145,56 g/mol
MDL Number: MFCD04972725
CAS nummer: 38186-84-4

Order Now

Specifikation testresultat

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Melting point 41.0 to 45.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR