trans-Crotononitrile ≥75.0% (contains ca. 20% cis- isomer)

Leverantör: TCI
Fara

Synonyms: trans-2-Butenenitrile

C3132-25ML C3132-100ML
TCIAC3132-25MLEA 445 SEK
TCIAC3132-25ML TCIAC3132-100ML
trans-Crotononitrile ≥75.0% (contains ca. 20% cis- isomer)
trans-Crotononitrile
Formel: C₄H₅N
Flampunkt: 16°C
CAS nummer: 627-26-9
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82fae095be3b5ef6f53418494adc3f54d848e9bb&vid=24bd2d8541471a38143c382311d565a80672f48c&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 75.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR