Clinofibrate ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 2,2'-((Cyclohexane-1,1-diylbis(4,1-phenylene))bis(oxy))bis(2-methylbutanoic acid), 2,2'-(4,4'-Cyclohexylidenediphenoxy)-2,2'-dimethyldibutanoic acid

C3100-200MG C3100-1G
TCIAC3100-200MGEA 990 SEK
TCIAC3100-200MG TCIAC3100-1G
Clinofibrate ≥98.0%
Clinofibrate
Formel: C₂₈H₃₆O₆
MW: 468,59 g/mol
MDL Number: MFCD00865769
CAS nummer: 30299-08-2
Merck Index: 14,02357

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 142.0 to 146.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR