7-Chloroquinoline ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
C3085-5G
C3085-25G
TCIAC3085-5GEA
1925
SEK
TCIAC3085-5G
TCIAC3085-25G
7-Chloroquinoline ≥98.0%
7-Chloroquinoline
Formel:
C₉H₆ClN MW: 163,6036 g/mol Kokpunkt: 268°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00956370 CAS nummer: 612-61-3 |
Specifikation testresultat
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...