5-Chloro-1-(4-piperidinyl)-2-benzimidazolidinone ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 5-Chloro-1-(piperidin-4-yl)-1H-benzo[d]imidazol-2(3H)-one, 5-Chloro-1-(4-piperidinyl)-2-benzimidazolinone

C3068-5G C3068-25G
TCIAC3068-5GEA 365 SEK
TCIAC3068-5G TCIAC3068-25G
5-Chloro-1-(4-piperidinyl)-2-benzimidazolidinone ≥98.0% (by GC, titration analysis)
5-Chloro-1-(4-piperidinyl)-2-benzimidazolidinone
Formel: C₁₂H₁₄ClN₃O
MW: 251,72 g/mol
MDL Number: MFCD02093793
CAS nummer: 53786-28-0
EINECS: 258-771-6

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 226.0 to 230.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR