4-Chloro-8-(trifluoromethyl)quinoline ≥98.0%

Leverantör: TCI EUROPE
Varning

C3067-1G C3067-5G
TCIAC3067-1GEA 1400 SEK
TCIAC3067-1G TCIAC3067-5G
4-Chloro-8-(trifluoromethyl)quinoline ≥98.0%
4-Chloro-8-(trifluoromethyl)quinoline
Formel: C₁₀H₅ClF₃N
MW: 231,61 g/mol
MDL Number: MFCD00134578
CAS nummer: 23779-97-7
EINECS: 245-880-9

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 78.0 to 82.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR