4-Chloro-8-fluoroquinoline ≥98.0%

Leverantör: TCI
Varning

C3062-1G C3062-5G
TCIAC3062-1GEA 1225 SEK
TCIAC3062-1G TCIAC3062-5G
4-Chloro-8-fluoroquinoline ≥98.0%
4-Chloro-8-fluoroquinoline
Formel: C₉H₅ClFN
MDL Number: MFCD00278785
CAS nummer: 63010-72-0

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 97.0 to 101.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR