2-Cyano-6-methylpyridine ≥98.0%

Leverantör: TCI
Fara

Synonyms: 6-Methylpicolinonitrile, 6-Methyl-2-pyridinecarbonitrile, 6-Cyano-2-picoline, 6-Methylpyridine-2-carbonitrile

C3025-1G C3025-5G
TCIAC3025-1GEA 365 SEK
TCIAC3025-1G TCIAC3025-5G
2-Cyano-6-methylpyridine ≥98.0%
2-Cyano-6-methylpyridine
Formel: C₇H₆N₂
Kokpunkt: 120°C
MDL Number: MFCD00190586
CAS nummer: 1620-75-3
EINECS: 216-588-9
UN: 3439
ADR: 6.1,III

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 70.0 to 74.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR