5-Chloropicolinonitrile ≥98.0%
Leverantör: TCI EUROPE
Fara
Synonyms: 5-Chloro-2-cyanopyridine
C3023-5G C3023-25G
TCIAC3023-5GEA 575 SEK
TCIAC3023-5G TCIAC3023-25G
5-Chloropicolinonitrile ≥98.0%
5-Chloropicolinonitrile
Formel: C6H3ClN2
MW: 138,55 g/mol
Kokpunkt: 110°C
MDL Number: MFCD03788835
CAS nummer: 89809-64-3
UN: 3439
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 107.0 to 111.0 deg-C