2-Chloro-N-methylacetamide ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
C3014-5G
C3014-25G
TCIAC3014-5GEA
750
SEK
TCIAC3014-5G
TCIAC3014-25G
2-Chloro-N-methylacetamide ≥98.0%
2-Chloro-N-methylacetamide
Formel:
C₃H₆ClNO MW: 107,54 g/mol Kokpunkt: 116°C |
MDL Number:
MFCD00018913 CAS nummer: 96-30-0 EINECS: 202-497-1 |
Specifikation testresultat
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Melting point | 42.0 to 46.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...