3-Chloro-1,2,4-triazole ≥98.0%

Leverantör: TCI EUROPE
Varning

C3013-1G C3013-5G
TCIAC3013-1GEA 845 SEK
TCIAC3013-1G TCIAC3013-5G
3-Chloro-1,2,4-triazole ≥98.0%
3-Chloro-1,2,4-triazole
Formel: C₂H₂ClN₃
MW: 103,51 g/mol
MDL Number: MFCD00128812
CAS nummer: 6818-99-1
EINECS: 229-892-1

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 167.0 to 171.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR