2-Chloro-5-(trifluoromethyl)-4-iodopyridine ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 2-Chloro-4-iodo-5-(trifluoromethyl)pyridine
C2999-1G C2999-5G
TCIAC2999-1GEA 405 SEK
TCIAC2999-1G TCIAC2999-5G
2-Chloro-5-(trifluoromethyl)-4-iodopyridine ≥98.0%
2-Chloro-5-(trifluoromethyl)-4-iodopyridine
Formel: C₆H₂ClF₃IN
MW: 307,44 g/mol
MDL Number: MFCD01862099
CAS nummer: 505084-55-9
Order Now
Specifikation testresultat
Appearance White to Green to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 126.0 to 130.0 deg-C