5-Chloroquinoline ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

C2997-1G C2997-5G
TCIAC2997-1GEA 695 SEK
TCIAC2997-1G TCIAC2997-5G
5-Chloroquinoline ≥98.0% (by GC, titration analysis)
5-Chloroquinoline
Formel: C₉H₆ClN
MW: 163,61 g/mol
Kokpunkt: 263°C
Storage Temperature: Kylskåp
MDL Number: MFCD00239413
CAS nummer: 635-27-8

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR