2-Cyanobenzeneboronic acid-1,3-propanediol ester ≥97.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 2-(1,3,2-Dioxaborinan-2-yl)benzonitrile, 2-Cyanobenzeneboronic acid 1,3-Propanediol ester, 2-Cyanophenylboronic acid 1,3-Propanediol ester
C2993-1G C2993-5G
TCIAC2993-1GEA 470 SEK
TCIAC2993-1G TCIAC2993-5G
2-Cyanobenzeneboronic acid-1,3-propanediol ester ≥97.0%
2-Cyanobenzeneboronic acid-1,3-propanediol ester
Formel: C₁₀H₁₀BNO₂
MW: 187,00 g/mol
MDL Number: MFCD04039012
CAS nummer: 172732-52-4
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 47.0 to 51.0 deg-C