4-Chloro-5-methoxypyridazin-3(2H)-one ≥98.0% (by GC, titration analysis)

Leverantör: TCI
Varning

Synonyms: 4-Chloro-5-methoxy-3(2H)-pyridazinone

C2972-200MG C2972-1G
TCIAC2972-200MGEA 615 SEK
TCIAC2972-200MG TCIAC2972-1G
4-Chloro-5-methoxypyridazin-3(2H)-one ≥98.0% (by GC, titration analysis)
4-Chloro-5-methoxypyridazin-3(2H)-one
Formel: C₅H₅ClN₂O₂
MW: 160.56 g/mol
MDL Number: MFCD04971392
CAS nummer: 63910-43-0

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR