3-(9H-Carbazol-9-yl)phenylboronic acid (contains varying amounts of Anhydride)

Leverantör: TCI
Varning

Synonyms: 3-(9-Carbazolyl)benzeneboronic acid, 3-(9H-Carbazol-9-yl)benzeneboronic Acid, (3-(9H-Carbazol-9-yl)phenyl)boronic acid

C2967-1G C2967-5G
TCIAC2967-1GEA 415 SEK
TCIAC2967-1G TCIAC2967-5G
3-(9H-Carbazol-9-yl)phenylboronic acid (contains varying amounts of Anhydride)
3-(9H-Carbazol-9-yl)phenylboronic acid
Formel: C₁₈H₁₄BNO₂
MW: 287.13 g/mol
MDL Number: MFCD18072506
CAS nummer: 864377-33-3

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 107.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR