1-Cyclohexyl-2-thiourea 98,0 HPLC, N

Leverantör: TCI
Varning

Synonyms: Cyclohexylthiourea, 1-Cyclohexylthiourea

C2966-200MG C2966-1G
TCIAC2966-200MGEA 630 SEK
TCIAC2966-200MG TCIAC2966-1G
1-Cyclohexyl-2-thiourea 98,0 HPLC, N
1-Cyclohexyl-2-thiourea
Formel: C₇H₁₄N₂S
MW: 158.27 g/mol
MDL Number: MFCD00060341
CAS nummer: 5055-72-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6feef0e8c370adde803c5d11fee28c0a0f5431db&vid=6feef0e8c370adde803c5d11fee28c0a0f5431db&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 162.0 to 166.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR