(4-Chloro-2-pyridinyl)methanol ≥98.0%

Leverantör: TCI EUROPE

Synonyms: (4-Chloropyridin-2-yl)methanol, 4-Chloro-2-(hydroxymethyl)pyridine

C2950-1G C2950-5G
TCIAC2950-1GEA 415 SEK
TCIAC2950-1G TCIAC2950-5G
(4-Chloro-2-pyridinyl)methanol ≥98.0%
(4-Chloro-2-pyridinyl)methanol
Formel: C₆H₆ClNO
MW: 143,5709 g/mol
Kokpunkt: 66°C
MDL Number: MFCD07437885
CAS nummer: 63071-10-3

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 43.0 to 47.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR