1-(Chloroacetyl)pyrrolidine ≥97.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 2-Chloro-1-(pyrrolidin-1-yl)ethanone
C2948-1G C2948-5G
TCIAC2948-1GEA 685 SEK
TCIAC2948-1G TCIAC2948-5G
1-(Chloroacetyl)pyrrolidine ≥97.0%
1-(Chloroacetyl)pyrrolidine
Formel: C₆H₁₀ClNO
MW: 147,60 g/mol
Kokpunkt: 105°C
MDL Number: MFCD01217237
CAS nummer: 20266-00-6
EINECS: 243-658-6
Order Now
Specifikation testresultat
Appearance White to Orange to Green powder to lump
Purity (GC) min. 97.0 %
Melting point 44.0 to 48.0 deg-C