2-Chloro-5-(trifluoromethyl)phenol ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 4-Chloro-3-hydroxybenzotrifluoride, 1-Chlor-2-hydroxy-4-(trifluormethyl)benzol

C2947-5G C2947-25G
TCIAC2947-5GEA 455 SEK
TCIAC2947-5G TCIAC2947-25G
2-Chloro-5-(trifluoromethyl)phenol ≥98.0%
2-Chloro-5-(trifluoromethyl)phenol
Formel: C₇H₄ClF₃O
MW: 196,56 g/mol
Kokpunkt: 88°C
Flampunkt: 46°C
MDL Number: MFCD00019995
CAS nummer: 40889-91-6
UN: 1993
ADR: 3,III

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR