3-Chloro-2-fluorophenylboronic acid (contains varying amounts of Anhydride)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
(3-Chloro-2-fluorophenyl)metaboric acid, Dihydroxy(3-chloro-2-fluorophenyl)borane, 3-Chloro-2-fluorophenylboronic Acid, 3-Chloro-2-fluorobenzeneboronic acid
C2946-1G
C2946-5G
TCIAC2946-1GEA
375
SEK
TCIAC2946-1G
TCIAC2946-5G
3-Chloro-2-fluorophenylboronic acid (contains varying amounts of Anhydride)
3-Chloro-2-fluorophenylboronic acid
Formel:
C₆H₅BClFO₂ MW: 174,37 g/mol Smältpunkt: 247°C |
MDL Number:
MFCD05664224 CAS nummer: 352535-82-1 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 112.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...