1,2,3,4-Cyclopentanetetracarboxylic dianhydride ≥99.0%, purified by sublimation

Leverantör: TCI
Varning

C2920-1G C2920-5G
TCIAC2920-1GEA 615 SEK
TCIAC2920-1G TCIAC2920-5G
1,2,3,4-Cyclopentanetetracarboxylic dianhydride ≥99.0%, purified by sublimation
1,2,3,4-Cyclopentanetetracarboxylic dianhydride
Formel: C₉H₆O₆
MW: 210.14 g/mol
MDL Number: MFCD00046872
CAS nummer: 6053-68-5
EINECS: 227-964-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=962ada1a1c8ec5ca392f41ab13064756db55b15b&vid=b85f44d0f9196795f0c0207e374a67a30265620b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White powder to crystal
Purity (Neutralization titration) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR