3-Chloro-5-fluorophenylboronic acid (contains varying amounts of Anhydride)

Leverantör: TCI EUROPE
Varning

Synonyms: 3-Chloro-5-fluorobenzeneboronic acid, 3-Chloro-5-fluorophenlyboronic acid

C2914-1G C2914-5G
TCIAC2914-1GEA 470 SEK
TCIAC2914-1G TCIAC2914-5G
3-Chloro-5-fluorophenylboronic acid (contains varying amounts of Anhydride)
3-Chloro-5-fluorophenylboronic acid
Formel: C₆H₅BClFO₂
MW: 174,37 g/mol
MDL Number: MFCD06801741
CAS nummer: 328956-61-2

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR