4-Chloro-3-nitrobenzophenone ≥98.0%

Leverantör: TCI EUROPE
Varning

C2906-5G C2906-25G
TCIAC2906-5GEA 270 SEK
TCIAC2906-5G TCIAC2906-25G
4-Chloro-3-nitrobenzophenone ≥98.0%
4-Chloro-3-nitrobenzophenone
Formel: C₁₃H₈ClNO₃
MW: 261,66 g/mol
Kokpunkt: 235°C
MDL Number: MFCD00007081
CAS nummer: 56107-02-9
EINECS: 259-995-7

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 104.0 to 108.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR