3-Chloro-1-phenylpropan-1-ol ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 3-Chloro-1-phenyl-1-propanol, 3-Chloro-1-phenyl-propanol, alpha-(2-Chloroethyl)benzyl alcohol

C2904-5G C2904-25G
TCIAC2904-5GEA 415 SEK
TCIAC2904-5G TCIAC2904-25G
3-Chloro-1-phenylpropan-1-ol ≥98.0%
3-Chloro-1-phenylpropan-1-ol
Formel: C9H11ClO
MW: 170,6400 g/mol
Kokpunkt: 116°C
Storage Temperature: Kylskåp
MDL Number: MFCD09878800
CAS nummer: 18776-12-0

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 31.0 to 36.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR